ContactUsLinkComponent
Agarose Gel Cup 1% TBE without stain. Includes ten Gel Cups with preweighed 1% agarose and 1 bottle of 100ml Tris-Borate-EDTA (TBE) buffer concentrate. 1 * 10 items
Catalog # EMBIM3151TBE
Supplier:  EMBITEC
Agarose Gel Cup 1% TBE without stain. Includes ten Gel Cups with preweighed 1% agarose and 1 bottle of 100ml Tris-Borate-EDTA (TBE) buffer concentrate. 1 * 10 items
Catalog # EMBIM3151TBE
Supplier:  EMBITEC
Frequently Bought Together