ContactUsLinkComponent
MiniLab Candy colour inquiry. Includes ten 1% agarose gel cups, one bottle of 100ml Tris-Borate-EDTA (TBE) buffer concentrate, one b ag of 1,7ml microcentrifuge tubes, one bag of 1 – 200µl micropipette tips, dye extraction buffer, candies in six colo 1 * 1 KIT
Catalog # EMBIM3009
Supplier:  EMBITEC
MiniLab Candy colour inquiry. Includes ten 1% agarose gel cups, one bottle of 100ml Tris-Borate-EDTA (TBE) buffer concentrate, one b ag of 1,7ml microcentrifuge tubes, one bag of 1 – 200µl micropipette tips, dye extraction buffer, candies in six colo 1 * 1 KIT
Catalog # EMBIM3009
Supplier:  EMBITEC
Frequently Bought Together