ContactUsLinkComponent
DNA marker universal MiniOne. Supplied in ready-to-use format and consists of 9 double-strand DNA fragments with sizes of 10K-6K-3K- 2K-1K-800-600-400-200 base pairs. (50 loads) 1 * 500 µl
Catalog # EMBIM3144
Supplier:  EMBITEC
DNA marker universal MiniOne. Supplied in ready-to-use format and consists of 9 double-strand DNA fragments with sizes of 10K-6K-3K- 2K-1K-800-600-400-200 base pairs. (50 loads) 1 * 500 µl
Catalog # EMBIM3144
Supplier:  EMBITEC
Frequently Bought Together