DNA ladder 1kb. Ready to load DNA marker contains 15 DNA fragments ranging from 1kb to 15kb with Xylene Cyanol FF and Orange G track ing dyes. (for 80 to 100 loads) 1 * 1.000 µl
Catalog # EMBIM3116
Supplier: EMBITEC
DNA ladder 1kb. Ready to load DNA marker contains 15 DNA fragments ranging from 1kb to 15kb with Xylene Cyanol FF and Orange G track ing dyes. (for 80 to 100 loads) 1 * 1.000 µl
Catalog # EMBIM3116
Supplier: EMBITEC
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, DNA ladders
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Polyvinylpyrrolidone, M.W. ∼ 58,000 (K29-32)
Supplier: Thermo Fisher Scientific