Set MiniOne micropipette with case. Includes variable pipette 2-20µl (M2008), variable pipette 20-200µl (M2010) and variable pipette 100-1000 µl (M2011). 1 * 1 SET
Katalognr. EMBIM2016
Anbieter: EMBITEC
Set MiniOne micropipette with case. Includes variable pipette 2-20µl (M2008), variable pipette 20-200µl (M2010) and variable pipette 100-1000 µl (M2011). 1 * 1 SET
Katalognr. EMBIM2016
Anbieter: EMBITEC
Frequently Bought Together
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Anbieter: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Anbieter: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Anbieter: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Anbieter: VWR Chemicals