ContactUsLinkComponent
405 Target Protein,,MPRA Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IF/ICC,ELISA Recommended Dilution,,WB 1:500~1:1000, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse Host Species,,Rabbit Immunogen, 1 * 100 µG
Catalog # GENOITA5863100U405
Supplier:  G-Biosciences
405 Target Protein,,MPRA Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IF/ICC,ELISA Recommended Dilution,,WB 1:500~1:1000, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse Host Species,,Rabbit Immunogen, 1 * 100 µG
Catalog # GENOITA5863100U405
Supplier:  G-Biosciences
Frequently Bought Together