DHR 123 PROBE HESE THEN LEAD TO OXIDATIVE STRESS. ROS INDUCED DAMAGE IN DNA, PROTEIN AND LIPIDS CAN LEAD TO IMPORTANT CONSEQUENCES IN CELLS. AFTER CELL UPTAKE, DHR 123 IS OXIDIZED BY ROS INTO A FLUORESCENT COMPOUND. 1 * 1 KIT
Catalog # GENOBAQ044
Supplier: G-Biosciences
Frequently Bought Together