ContactUsLinkComponent
Mitosis, l.s. from Hyacinthus root tips showing all stages of plant mitosis carefully stained with a quadruple stain. Specially large chromosomes, for demonstration of plant mitosis 1 * 1 items
Catalog # LIEDAS1166E
Anbieter:  LIEDER JOHANNES
Mitosis, l.s. from Hyacinthus root tips showing all stages of plant mitosis carefully stained with a quadruple stain. Specially large chromosomes, for demonstration of plant mitosis 1 * 1 items
Catalog # LIEDAS1166E
Anbieter:  LIEDER JOHANNES
Frequently Bought Together