ContactUsLinkComponent
Chemistry pumping unit PC 3002 VARIO EKP o. C., without controller, with Peltronic exhaust vapor condenser, 200-230 V / 50-60 Hz without mains cable 1 * 1 items
Catalog # VACU22614242
Supplier:  VACUUBRAND
Chemistry pumping unit PC 3002 VARIO EKP o. C., without controller, with Peltronic exhaust vapor condenser, 200-230 V / 50-60 Hz without mains cable 1 * 1 items
Catalog # VACU22614242
Supplier:  VACUUBRAND
Frequently Bought Together