ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Plastin L/LCP1 detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, Direct ELISA in Human Mouse Rat . 1 * 100 µG
Catalog # BSBTA03361
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Plastin L/LCP1 detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, Direct ELISA in Human Mouse Rat . 1 * 100 µG
Catalog # BSBTA03361
Supplier:  Boster Bio
Frequently Bought Together