Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Hydrobromic acid 47%, GPR RECTAPUR®

25 L
  VWRC20208.460
 
 
Hydrobromic acid 47%, GPR RECTAPUR®
  VWRC20208.460
 :  10035-10-6
Hydrobromic acid 47%, GPR RECTAPUR®
Hydrobromic acid 47%, GPR RECTAPUR®

25 L

 :  10035-10-6
 
SFr. 4’560.00
   
1 * 25 L
 
 :  Hydrogen bromide
  •  :10035-10-6
  •  :HBr
  •  :MFCD00011323
  •  :80.91 g/mol
  •  :1.47 g/cm³ (20 °C)
  •  :Ambient
  •  :233-113-0
  •  :1788
  •  :8,II
 

 

  • Assay:
    Min. 47 %
  • Phosphate, Phosphite (as PO4):
    max 0.002%
  • Sulphate and sulphite (as SO4):
    max 0.005%
  • Heavy metals (as Pb):
    max 0.001%
  • Fe (Iron):
    max 0.0005%

 

  • Pk:
    25 L

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.