L-KYNURENINE SULFATE (RING-D4, 3,3-D2, 97%+
Catalog # CAMBDLM-7842.5MG
Supplier: CAMBRIDGE ISOTOPE LABS
L-KYNURENINE SULFATE (RING-D4, 3,3-D2, 97%+
Catalog # CAMBDLM-7842.5MG
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Syringe filters, prefilter, sterile, GD/X™, Whatman™
Supplier: Whatman products (Cytiva)
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
2-Methylbutane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
3-HYDROXY-DL-KYNURENINE 1 * 25 mg
Supplier: Cayman Chemical