: 3-AMINO-1-METHYLCYCLOHEXAN-1-O
Catalog # UKROBBV-43557729.1
Marketsource
Supplier: UKRORGSYNTHESIS
: 3-AMINO-1-METHYLCYCLOHEXAN-1-O
Catalog # UKROBBV-43557729.1
Supplier: UKRORGSYNTHESIS
Marketsource
CAS Number:
: 3-AMINO-1-METHYLCYCLOHEXAN-1-O
CAS Number:
Product Details & Documents
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
#: 1.00030.2500
Supplier: Merck
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
#: 1.09439.1000
Supplier: Merck



