ContactUsLinkComponent
BB 250 XL | 3-N 400 V, 50 Hz wc-hardened steel Complete with hopper door, collector and chassis Initial particle size <120 x 90 mm Final fineness <2 mm depending on the sample Opening of the jaws: 120 x 90 mm Easy cleaning thanks to the front door Ov 1 * 1 items
Catalog # RETS21.001.0002
Supplier:  Retsch GmbH
BB 250 XL | 3-N 400 V, 50 Hz wc-hardened steel Complete with hopper door, collector and chassis Initial particle size <120 x 90 mm Final fineness <2 mm depending on the sample Opening of the jaws: 120 x 90 mm Easy cleaning thanks to the front door Ov 1 * 1 items
Catalog # RETS21.001.0002
Supplier:  Retsch GmbH
Frequently Bought Together