ContactUsLinkComponent
schuett phoenix II adapter for the connection of C206 butane cartridges, incl. reducing regulator and gas safety hose (0.5 m) 1 * 1 ST
Catalog # SCUT3357232
schuett phoenix II adapter for the connection of C206 butane cartridges, incl. reducing regulator and gas safety hose (0.5 m) 1 * 1 ST
Catalog # SCUT3357232
Frequently Bought Together