OMEPRAZOLE IMPURITY C 1 * 30 mg
Catalog # PHR1988-30MG
Supplier: Merck
Frequently Bought Together
Buffer solutions
Supplier: REAGECON
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Distillation standards for the petroleum industry, Certified Reference Materials
Supplier: VWR Chemicals
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
Aspartame, Reference Standards
Supplier: USP
N-Nitroso-di-iso-propylamine 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
REAGECON PH 1.68 BUFFER SOLUTION AT 25C 1 * 1 L
Supplier: REAGECON
Propranolol Hydrochloride, Reference Standard
Supplier: USP
STANDARD S-21+K-LI-SB 100PPM 1 * 200 g
Supplier: LGC Standards PROMOCHEM
2,6-Diaminotoluene
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, γ-Tocotrienol
Supplier: Merck
Hydrochloric acid 1 N USP Test Solution (TS)
Supplier: VWR Chemicals
pH buffer solutions, colour coded
Supplier: Hach
Supelco® Organic Reference Standard, (+)-Carvone
Supplier: Merck