ContactUsLinkComponent
GL-Reductions EX, Black screw cap made of PPS-EX, movable reduction body made of PTFE-EX with o-ring made of FKM for transition to g lthreads. The body can be moved independently from the screw cap so that the completely assembled reduction can be re 1 * 1 items
Catalog # BOHLD872-20
Supplier:  Bohlender
undefined
GL-Reductions EX, Black screw cap made of PPS-EX, movable reduction body made of PTFE-EX with o-ring made of FKM for transition to g lthreads. The body can be moved independently from the screw cap so that the completely assembled reduction can be re 1 * 1 items
Catalog # BOHLD872-20
Supplier:  Bohlender
Frequently Bought Together