ContactUsLinkComponent
Multiple Distributors for Barrels, Screw cap made of glass-fibre reinforced PTFE for barrels with female thread, body with GL-threa ded necks made of PTFE. Very good chemical resistance, for working temperatures up to max. 1 * 1 items
Catalog # BOHLD698-06
Supplier:  Bohlender
undefined
Multiple Distributors for Barrels, Screw cap made of glass-fibre reinforced PTFE for barrels with female thread, body with GL-threa ded necks made of PTFE. Very good chemical resistance, for working temperatures up to max. 1 * 1 items
Catalog # BOHLD698-06
Supplier:  Bohlender
Frequently Bought Together