ContactUsLinkComponent
Screw In Stopper, For connection to BOLA Reactor Lids. For closure of non-used connectors. With hexagonal gripping surface in standa rd wrench size. 1 * 1 items
Catalog # BOHLB153-08
Supplier:  Bohlender
undefined
Screw In Stopper, For connection to BOLA Reactor Lids. For closure of non-used connectors. With hexagonal gripping surface in standa rd wrench size. 1 * 1 items
Catalog # BOHLB153-08
Supplier:  Bohlender
Frequently Bought Together