ContactUsLinkComponent
roll of 1000 m of creped filter paper, grade 55/N, 73g/m2, laize 600 m
Catalog # BINZ1.470.06001000
Supplier:  Ahlstrom-Munksjö
roll of 1000 m of creped filter paper, grade 55/N, 73g/m2, laize 600 m
Catalog # BINZ1.470.06001000
Supplier:  Ahlstrom-Munksjö
Frequently Bought Together