Densimeter ISO 649 L50 0.950-1.000 : 0.0005 g/ml Tp.20°C with certificate of 3-point calibration 1 * 1 items
ALLA3500LC09520BCA
Densimeter ISO 649 L50 0.950-1.000 : 0.0005 g/ml Tp.20°C with certificate of 3-point calibration 1 * 1 items
ALLA3500LC09520BCA
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection