Orders LinkContactUsLinkComponent
Autoclavable goggles
Autoclavable goggles
Cleanroom goggles, Anti-UV, anti-fog, autoclavable, directly vented, Clear PC, Blue
Catalog # 111-2428
Supplier:   UNIVET
Autoclavable goggles
Catalog # 111-2428
Supplier:  UNIVET
Autoclavable goggles
Autoclavable goggles
Cleanroom goggles, Anti-UV, anti-fog, autoclavable, directly vented, Clear PC, Blue
Price
SFr. 457.00
Log in for availability
1 * 5 ST
Product Details & Documents
The autoclavable goggles are designed for use in rooms with controlled contaminations or in aseptic conditions.
  • Available with direct or indirect ventilation
  • Elastic band in anti-static silicone with quick and smooth system
  • RX insert
  • Goggles with polycarbonate lens reusable up to 40 autoclave
The soft structure supports multiple cycles of steam, gamma or beta sterilisation without perceptible changes. The autoclavable goggles has a polycarbonate lens with anti-fog coating AF121 and anti-scratch.
Certifications: EN 166.
Delivery information: Available with correction insert and replacement lens.
Specifications
  • Type:
    Anti-UV, anti-fog, autoclavable, directly vented
  • Colour:
    Blue
  • Description:
    Cleanroom goggles
  • Seq:
    03
  • Lenses:
    Clear PC
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare lens for 611S, clear PC
    Spare lens for 611S, clear PC
    UNVT042.060
    Log in for availability
    1 * 1 ST - SFr. 19.00
Product Information
  • Catalog #
    Availability
    Price
  • Head band, silicone, 700×30×1,4 mm
    Head band, silicone, 700×30×1,4 mm
    UNVT3PA299
    Log in for availability
    1 * 1 ST - SFr. 25.20
Product Information
  • Catalog #
    Availability
    Price
  • RX optical insert for goggles
    RX optical insert for goggles
    UNVT601.OP.00.99
    Log in for availability
    1 * 1 ST - SFr. 18.30
Product Information
  • Catalog #
    Availability
    Price
  • Spare lens, clear PC
    Spare lens, clear PC
    UNVT042.120M
    Log in for availability
    1 * 1 ST - SFr. 42.80