ContactUsLinkComponent
HDPE collection can red with stainless steel fittings cap 7,5L 1 * 1 items
Catalog # DUEH5.147.62
Supplier:  DUPERTHAL
HDPE collection can red with stainless steel fittings cap 7,5L 1 * 1 items
Catalog # DUEH5.147.62
Supplier:  DUPERTHAL
Frequently Bought Together