ContactUsLinkComponent
NEW Compact laboratory safety shield: fits neatly around any stirrer hotplate to provide robust, convenient protection from splashes, aerosols and spills. Modified for back width 250mm, depth and other dimensions as standard model. 1 * 1 items
Catalog # ASYNADSLSSCUST1250
Supplier:  Asynt
NEW Compact laboratory safety shield: fits neatly around any stirrer hotplate to provide robust, convenient protection from splashes, aerosols and spills. Modified for back width 250mm, depth and other dimensions as standard model. 1 * 1 items
Catalog # ASYNADSLSSCUST1250
Supplier:  Asynt
Frequently Bought Together