ContactUsLinkComponent
5 components 1000mg / l each of Li N / A Mg Ca K in HNO3 2%. 1 * 1 items
  ACSDRM009989L1
 :  CPACHEM
5 components 1000mg / l each of Li N / A Mg Ca K in HNO3 2%. 1 * 1 items
  ACSDRM009989L1
 :  CPACHEM
Frequently Bought Together