ContactUsLinkComponent
Center piece individually without attachment Limit determination of arsenic NS 29/32, arsenic, complete 3-part with piston, attachme nt and tension springs, DAB 7 D. about 60mm x height about 270mm 1 * 1 items
Catalog # NEUB2390-020-29
Center piece individually without attachment Limit determination of arsenic NS 29/32, arsenic, complete 3-part with piston, attachme nt and tension springs, DAB 7 D. about 60mm x height about 270mm 1 * 1 items
Catalog # NEUB2390-020-29
Frequently Bought Together