KINETEX 1.7 µm EVO C18 100 Å, LC COLUMN 100 X 2.1 M
Catalog # PHEN00D-4726-AN
Supplier: PHENOMENEX
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals