Clip Cap PP 10 g/m² Comfort, blue, latex free, Made in France, in dispenser box 1 * 300 items
HOPE812102
: Medicom
Clip Cap PP 10 g/m² Comfort, blue, latex free, Made in France, in dispenser box 1 * 300 items
HOPE812102
: Medicom
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection