ContactUsLinkComponent
PurePlue® Deep Well Plates for Hamiliton Forensic Workstation. 96 wells. 2.0ml per well. With Barcode. Rnase/Dnase, ATP Free. Non-pyrogenic. DNA Free 10 plates per pack. 1 * 10 items
Catalog # 3905-520-020-9
Supplier:  LABCON
PurePlue® Deep Well Plates for Hamiliton Forensic Workstation. 96 wells. 2.0ml per well. With Barcode. Rnase/Dnase, ATP Free. Non-pyrogenic. DNA Free 10 plates per pack. 1 * 10 items
Catalog # 3905-520-020-9
Supplier:  LABCON
Frequently Bought Together