ContactUsLinkComponent
COD Complete System MD 100 Delivery Content Photometer, COD Reactor RD 125, COD tube tests 0 - 150 mg/l, 0 - 1500 mg/l , Tube stand, Instruction manual, Warranty information 1 * 1 SET
Catalog # LOVI276130
Supplier:  Lovibond
COD Complete System MD 100 Delivery Content Photometer, COD Reactor RD 125, COD tube tests 0 - 150 mg/l, 0 - 1500 mg/l , Tube stand, Instruction manual, Warranty information 1 * 1 SET
Catalog # LOVI276130
Supplier:  Lovibond
Frequently Bought Together