H-12-1.5-PLL: 12 mm diameter, # 1.5 thickness, 45 pieces of CE certified crystal clear German glass coverslips with poly-L-lysine coating on both sides for cell culture and confocal fluorescence microscopy, sealed 15 pieces per bag x 3 = 45, steriliz 1 * 45 ST
Catalog # MSPPH-12-1.5-PLL
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together