H-12-1.5-PDL: 12mm diameter, #1.5 thickness, 45 pieces of CE certified crystal clear German glass coverslips with poly-D-lysine coating on both sides for cell culture and confocal fluorescence microscopy, sealed 15 pieces per bag x 3 = 45, sterilized 1 * 45 ST
Catalog # MSPPH-12-1.5-PDL
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together