Masterflex®, Power Supply
Power supply, dual voltage
Catalog # 181-0527
Supplier: VWR Collection
Jump to:
Product Details & Documents
Specifications
- Description:Power supply, dual voltage
- Seq:03
- Cat. No.:181-0527
- Input:100 - 240 V
- Output:100/200 V