Orders LinkContactUsLinkComponent
Masterflex®, Power Supply
Masterflex®, Power Supply
Power supply, dual voltage
Catalog # 181-0525
Supplier:   VWR Collection
Masterflex®, Power Supply
Catalog # 181-0525
Supplier:  VWR Collection
Masterflex®, Power Supply
Power supply, dual voltage
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Power supply, dual voltage
  • Seq:
    01
  • Cat. No.:
    181-0525
  • Input:
    100 - 240 V
  • Output:
    100/200 V