CHLORDIOX PLUS CR-1 REPLACEMENT REAGENT 1 * 25 mL
Catalog # PALIPT546
Supplier: Palintest
Frequently Bought Together
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Manganese ≥99.3% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
CHLORDIOX PLUS CR-2 REPLACEMENT REAGENT 1 * 25 mL
Supplier: Palintest