S.O.C Medium is used in bacterial cell transformation after heat-shock treatment to the DNA-competent cell mixture to obtain maximal transformation efficiency. 1 * 50 mL
Catalog # GENO786-1280
Supplier: G-Biosciences
S.O.C Medium is used in bacterial cell transformation after heat-shock treatment to the DNA-competent cell mixture to obtain maximal transformation efficiency. 1 * 50 mL
Catalog # GENO786-1280
Supplier: G-Biosciences
Frequently Bought Together
Glass beads
Supplier: GLASWARENFABRIK KARL HECHT
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Electroporation Cuvettes
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF