ContactUsLinkComponent
Simple Plex Cartridge Kit for 32 samples, containing IL-13,IL-33,IL-4,IL-5 for use with Human Plasma / Serum. Includes wash buffer a nd sample diluent SD13. 1 * 1 items
Catalog # XXXXSPCKC-PS000166
Supplier:  Chemicals
Simple Plex Cartridge Kit for 32 samples, containing IL-13,IL-33,IL-4,IL-5 for use with Human Plasma / Serum. Includes wash buffer a nd sample diluent SD13. 1 * 1 items
Catalog # XXXXSPCKC-PS000166
Supplier:  Chemicals
Frequently Bought Together