ContactUsLinkComponent
GreenGel cup 0,6% TBE with GelGreen DNA stain. Re-meltable agarose gel with fluorescent DNA stain in TBE buffer for 0,8% gels. 1 * 10 items
Catalog # EMBIM3140TBE
Supplier:  EMBITEC
GreenGel cup 0,6% TBE with GelGreen DNA stain. Re-meltable agarose gel with fluorescent DNA stain in TBE buffer for 0,8% gels. 1 * 10 items
Catalog # EMBIM3140TBE
Supplier:  EMBITEC
Frequently Bought Together