ContactUsLinkComponent
MiniLab CSI forensics. Includes DNA Samples (5), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcentrifuge tubes (55 tubes ), micropipette tips (60 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3005
Supplier:  EMBITEC
MiniLab CSI forensics. Includes DNA Samples (5), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcentrifuge tubes (55 tubes ), micropipette tips (60 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3005
Supplier:  EMBITEC
Frequently Bought Together