ContactUsLinkComponent
MiniLab electrophoresis 101. Includes DNA and dye samples (6 samples), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcent rifuge tubes (66 tubes), micropipette tips (70 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3001
Supplier:  EMBITEC
MiniLab electrophoresis 101. Includes DNA and dye samples (6 samples), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcent rifuge tubes (66 tubes), micropipette tips (70 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3001
Supplier:  EMBITEC
Frequently Bought Together