AUSHON QUANTERIX SPX CIRAPLEX® CHEMILUMINESCENT ARRAY KIT: 3-PLEX HUMAN IL-4, IL-5, IL-13 (SERUM SAMPLES
Catalog # XXXX100-0454
Supplier: Chemicals
AUSHON QUANTERIX SPX CIRAPLEX® CHEMILUMINESCENT ARRAY KIT: 3-PLEX HUMAN IL-4, IL-5, IL-13 (SERUM SAMPLES
Catalog # XXXX100-0454
Supplier: Chemicals
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection