Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Magnetic sets

Magnetic sets

Cylindrical transfection magnets, sets of four, Ø 2 mm, 5 mm and 10 mm
  OZBIIV-MAG3
Magnetic sets
  OZBIIV-MAG3
Magnetic sets

Cylindrical transfection magnets, sets of four, Ø 2 mm, 5 mm and 10 mm

 
SFr. 64.51
   
1 * 1 ST
 

These magnetic sets have been developped for efficient in vivo transfection or transduction with in vivo Magnetofection technology.

  • Can be easily cleaned and decontaminated with 70% ethanol, or by dry or wet sterilisation
  • Can be used at room temperature, 37 °C, +4 °C, etc.
  • Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targetd site
  • Solid, completely reusable, it is a one-time buy

Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targeted site.

 
  • Description:
    Cylindrical transfection magnets, sets of four, Ø 2 mm, 5 mm and 10 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.