ContactUsLinkComponent
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, non-woven 52g/m² blue and green, 120x120cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1349
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, non-woven 52g/m² blue and green, 120x120cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1349
Frequently Bought Together