ContactUsLinkComponent
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, SMMS 60g/m² dark blue and non-woven 67g/m² green, 120x120cm, DIN EN ISO 11607 1 * 120 items
Catalog # 129-1344
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, SMMS 60g/m² dark blue and non-woven 67g/m² green, 120x120cm, DIN EN ISO 11607 1 * 120 items
Catalog # 129-1344
Frequently Bought Together