ContactUsLinkComponent
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, SSMMS 43g/m² light blue and non-woven 57g/m² green, 75x75cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1338
Stericlin® sterilisation wrap interleaved, alternately stacked sheets, SSMMS 43g/m² light blue and non-woven 57g/m² green, 75x75cm, DIN EN ISO 11607 1 * 200 items
Catalog # 129-1338
Frequently Bought Together