ContactUsLinkComponent
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SSMMS 55g/m² dark blue, inner sheet non-woven 67g/m² green, 100x100cm, DIN EN ISO 11607 1 * 80 items
Catalog # 129-1335
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SSMMS 55g/m² dark blue, inner sheet non-woven 67g/m² green, 100x100cm, DIN EN ISO 11607 1 * 80 items
Catalog # 129-1335
Frequently Bought Together