ContactUsLinkComponent
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 47g/m² light blue, inner sheet non-woven 57g/m² green, 120x120cm, DIN EN ISO 11607 1 * 72 items
Catalog # 129-1331
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 47g/m² light blue, inner sheet non-woven 57g/m² green, 120x120cm, DIN EN ISO 11607 1 * 72 items
Catalog # 129-1331
Frequently Bought Together