ContactUsLinkComponent
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 47g/m² light blue, inner sheet non-woven 57g/m² green, 90x90cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1329
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 47g/m² light blue, inner sheet non-woven 57g/m² green, 90x90cm, DIN EN ISO 11607 1 * 100 items
Catalog # 129-1329
Frequently Bought Together