ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 60g/m dark blue, inner sheet SSMMS 47g/m gree n, 100x100cm, DIN EN ISO 1160
Catalog # 129-1325
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 60g/m dark blue, inner sheet SSMMS 47g/m gree n, 100x100cm, DIN EN ISO 1160
Catalog # 129-1325