ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 60g/m dark blue, inner sheet SSMMS 47g/m gree n, 90x90cm, DIN EN ISO 1160
Catalog # 129-1324
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SMMS 60g/m dark blue, inner sheet SSMMS 47g/m gree n, 90x90cm, DIN EN ISO 1160
Catalog # 129-1324